The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001015008.1 | NP_001015008.1 | 10 - 11 | 1117 | 69 |
GenBank mRNA | BC091180.1 | - | 10 - 11 | 1117 | 69 |