Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dnali1
Entrez Gene ID
298524
Gene Name
dynein, axonemal, light intermediate chain 1
Gene Aliases
-
Chromosome Location
Chr.5: 143053365 - 143062203 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031647.1 NP_001026817.1 1 - 2 89 68
GenBank mRNA BC099087.1 - 1 - 2 112 68

View Full Product Details