The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031647.1 | NP_001026817.1 | 5 - 6 | 740 | 63 |
GenBank mRNA | BC099087.1 | - | 5 - 6 | 763 | 63 |