The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106701.2 | NP_001100171.1 | 3 - 4 | 393 | 79 |
GenBank mRNA | BC158867.1 | - | 3 - 4 | 393 | 79 |
FQ219403.1 | - | 2 - 3 | 800 | 79 |