Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam76b
Entrez Gene ID
367021
Gene Name
family with sequence similarity 76, member B
Gene Aliases
RGD1311077
Chromosome Location
Chr.8: 12355767 - 12378040 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108994.1 NP_001102464.1 5 - 6 812 136
XM_006242526.1 XP_006242588.1 5 - 6 797 136
XM_006242527.1 XP_006242589.1 5 - 6 797 136
XM_006242529.2 XP_006242591.1 5 - 6 797 136
XM_006242530.2 XP_006242592.1 5 - 6 797 136
GenBank mRNA FQ212292.1 - 5 - 6 507 136

View Full Product Details