Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Snx30
Entrez Gene ID
298033
Gene Name
sorting nexin family member 30
Gene Aliases
RGD1311367
Chromosome Location
Chr.5: 77052484 - 77157915 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106651.1 NP_001100121.1 7 - 8 1323 60

View Full Product Details