The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008868.1 | NP_001008868.1 | 1 - 2 | 75 | 65 |
XM_008762531.1 | XP_008760753.1 | 1 - 2 | 86 | 65 |