Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wfdc9
Entrez Gene ID
408228
Gene Name
WAP four-disulfide core domain 9
Gene Aliases
-
Chromosome Location
Chr.3: 161112251 - 161115324 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008762531.1 XP_008760753.1 2 - 3 227 85

View Full Product Details