Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tubd1
Entrez Gene ID
287592
Gene Name
tubulin, delta 1
Gene Aliases
-
Chromosome Location
Chr.10: 73865828 - 73886976 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105826.1 NP_001099296.1 1 - 2 41 81

View Full Product Details