Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prpsap1
Entrez Gene ID
64390
Gene Name
phosphoribosyl pyrophosphate synthetase-associated protein 1
Gene Aliases
-
Chromosome Location
Chr.10: 105430516 - 105457942 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022545.3 NP_071990.2 4 - 5 627 120
XM_008768391.1 XP_008766613.1 4 - 5 564 120
GenBank mRNA BC078822.1 - 4 - 5 602 120
D26073.1 - 4 - 5 525 120

View Full Product Details