The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013973.1 | NP_001013995.1 | 11 - 12 | 2577 | 61 |
XM_006247579.2 | XP_006247641.1 | 11 - 12 | 2745 | 61 | |
XM_006247580.2 | XP_006247642.1 | 12 - 13 | 2864 | 61 | |
GenBank mRNA | AB552845.1 | - | 9 - 10 | 2322 | 61 |
BC083567.1 | - | 11 - 12 | 2577 | 61 |