Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prss22
Entrez Gene ID
302971
Gene Name
protease, serine, 22
Gene Aliases
-
Chromosome Location
Chr.10: 13370098 - 13374808 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106984.1 NP_001100454.1 5 - 6 795 80
GenBank mRNA AJ005642.1 - 4 - 5 779 80

View Full Product Details