Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sh3pxd2b
Entrez Gene ID
303021
Gene Name
SH3 and PX domains 2B
Gene Aliases
RGD1309926
Chromosome Location
Chr.10: 17209152 - 17296449 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006246109.2 XP_006246171.1 4 - 5 370 85
XM_008767621.1 XP_008765843.1 3 - 4 941 85
XM_008767622.1 XP_008765844.1 3 - 4 941 85

View Full Product Details