Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
March10
Entrez Gene ID
303596
Gene Name
membrane-associated ring finger (C3HC4) 10, E3 ubiquitin protein ligase
Gene Aliases
MARCH-X, RGD1311692, Rnf190
Chromosome Location
Chr.10: 93591187 - 93681486 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013973.1 NP_001013995.1 4 - 5 456 83
XM_006247579.2 XP_006247641.1 4 - 5 624 83
XM_006247580.2 XP_006247642.1 5 - 6 743 83
XM_006247585.2 XP_006247647.1 4 - 5 601 83
GenBank mRNA AB552845.1 - 2 - 3 201 83
AB552846.1 - 2 - 3 201 83
BC083567.1 - 4 - 5 456 83

View Full Product Details