Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Med7
Entrez Gene ID
100911235
Gene Name
mediator complex subunit 7
Gene Aliases
Crsp9
Chromosome Location
Chr.10: 31532194 - 31536405 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006246161.1 XP_006246223.1 1 - 2 100 78

View Full Product Details