Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Narf
Entrez Gene ID
360681
Gene Name
nuclear prelamin A recognition factor
Gene Aliases
RGD1310894
Chromosome Location
Chr.10: 110445764 - 110463735 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039207.2 NP_001034296.1 10 - 11 1223 71
GenBank mRNA BC110050.1 - 10 - 11 1223 71

View Full Product Details