Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
B3gntl1
Entrez Gene ID
367384
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase-like 1
Gene Aliases
-
Chromosome Location
Chr.10: 110799406 - 110863071 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015035.1 NP_001015035.1 4 - 5 433 80
XM_006247904.2 XP_006247966.1 4 - 5 483 80
XM_008768503.1 XP_008766725.1 4 - 5 483 80
XM_008768504.1 XP_008766726.1 4 - 5 483 80
XM_008768505.1 XP_008766727.1 4 - 5 483 80
XM_008768506.1 XP_008766728.1 4 - 5 483 80
XM_008768507.1 XP_008766729.1 4 - 5 483 80
XM_008768508.1 XP_008766730.1 4 - 5 483 80
XM_008768509.1 XP_008766731.1 4 - 5 483 80
GenBank mRNA AY634365.1 - 4 - 5 433 80
BC111711.1 - 4 - 5 407 80

View Full Product Details