The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135158.1 | NP_001128630.1 | 27 - 28 | 3842 | 61 |
XM_008767787.1 | XP_008766009.1 | 27 - 28 | 3852 | 61 | |
GenBank mRNA | S68736.1 | - | 3 - 4 | 597 | 61 |