Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Spem1
Entrez Gene ID
691981
Gene Name
spermatid maturation 1
Gene Aliases
-
Chromosome Location
Chr.10: 56428447 - 56429748 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109653.1 NP_001103123.1 1 - 2 230 143

View Full Product Details