The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127522.1 | NP_001120994.1 | 1 - 2 | 379 | 59 |
XM_006247711.2 | XP_006247773.1 | 1 - 2 | 496 | 59 | |
GenBank mRNA | BC166454.1 | - | 1 - 2 | 353 | 59 |