The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039377.1 | NP_001034466.1 | 8 - 9 | 1219 | 72 |
GenBank mRNA | AY903231.1 | - | 8 - 9 | 1219 | 72 |
AY903232.1 | - | 8 - 9 | 1219 | 72 | |
AY903233.1 | - | 8 - 9 | 1219 | 72 |