Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Alox12b
Entrez Gene ID
287425
Gene Name
arachidonate 12-lipoxygenase, 12R type
Gene Aliases
-
Chromosome Location
Chr.10: 55744646 - 55756524 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039377.1 NP_001034466.1 13 - 14 1900 60
GenBank mRNA AY903231.1 - 13 - 14 1900 60
AY903232.1 - 13 - 14 1900 60
AY903233.1 - 13 - 14 1900 60

View Full Product Details