Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bzrap1
Entrez Gene ID
287609
Gene Name
benzodiazepine receptor (peripheral) associated protein 1
Gene Aliases
-
Chromosome Location
Chr.10: 75054070 - 75078571 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008768165.1 XP_008766387.1 22 - 23 4808 100
XM_213427.7 XP_213427.5 22 - 23 4808 100

View Full Product Details