Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slfn2
Entrez Gene ID
303380
Gene Name
schlafen 2
Gene Aliases
-
Chromosome Location
Chr.10: 70370381 - 70376718 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107031.1 NP_001100501.1 1 - 2 242 71

View Full Product Details