Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wnt9b
Entrez Gene ID
303586
Gene Name
wingless-type MMTV integration site family, member 9B
Gene Aliases
-
Chromosome Location
Chr.10: 91785893 - 91807937 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107055.1 NP_001100525.1 1 - 2 136 122

View Full Product Details