Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prr15l
Entrez Gene ID
688163
Gene Name
proline rich 15-like
Gene Aliases
-
Chromosome Location
Chr.10: 84866786 - 84873434 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003750919.3 XP_003750967.1 1 - 2 249 68

View Full Product Details