Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sp2
Entrez Gene ID
303499
Gene Name
Sp2 transcription factor
Gene Aliases
-
Chromosome Location
Chr.10: 84890930 - 84920886 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107045.1 NP_001100515.1 6 - 7 1809 80
XM_006247303.2 XP_006247365.1 5 - 6 1571 80
GenBank mRNA BC168669.1 - 5 - 6 1577 80

View Full Product Details