Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zscan10
Entrez Gene ID
302962
Gene Name
zinc finger and SCAN domain containing 10
Gene Aliases
Zfp206, Znf206
Chromosome Location
Chr.10: 12929471 - 12939439 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006245870.2 XP_006245932.1 1 - 2 169 78

View Full Product Details