Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Smtnl2
Entrez Gene ID
679629
Gene Name
smoothelin-like 2
Gene Aliases
-
Chromosome Location
Chr.10: 58951845 - 58973020 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190998.1 NP_001177927.1 6 - 7 1156 80

View Full Product Details