Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Alox12e
Entrez Gene ID
303252
Gene Name
arachidonate 12-lipoxygenase, epidermal
Gene Aliases
12-LOX
Chromosome Location
Chr.10: 56927676 - 56935141 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107014.1 NP_001100484.1 10 - 11 1487 88

View Full Product Details