Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Alox12e
Entrez Gene ID
303252
Gene Name
arachidonate 12-lipoxygenase, epidermal
Gene Aliases
12-LOX
Chromosome Location
Chr.10: 56927676 - 56935141 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107014.1 NP_001100484.1 6 - 7 880 66

View Full Product Details