Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tmc8
Entrez Gene ID
688276
Gene Name
transmembrane channel-like 8
Gene Aliases
-
Chromosome Location
Chr.10: 106784827 - 106795307 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126301.1 NP_001119773.1 4 - 5 639 130
XM_006247818.2 XP_006247880.1 4 - 5 1087 130
XM_006247821.2 XP_006247883.1 4 - 5 621 130
XM_008768392.1 XP_008766614.1 4 - 5 612 130
GenBank mRNA BC160873.1 - 4 - 5 639 130

View Full Product Details