Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dnajc7
Entrez Gene ID
303536
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 7
Gene Aliases
CCRP
Chromosome Location
Chr.10: 88498271 - 88533747 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213625.1 NP_998790.1 2 - 3 164 143
XM_006247311.2 XP_006247373.1 3 - 4 525 143
GenBank mRNA AB178470.1 - 2 - 3 164 143
FQ212948.1 - 2 - 3 195 143

View Full Product Details