The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164543.1 | NP_001158015.1 | 1 - 2 | 306 | 86 |
GenBank mRNA | BC158725.1 | - | 1 - 2 | 306 | 86 |
FQ220528.1 | - | 1 - 2 | 70 | 86 |