Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ubtd2
Entrez Gene ID
287178
Gene Name
ubiquitin domain containing 2
Gene Aliases
RGD1305506
Chromosome Location
Chr.10: 17327100 - 17392432 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008767629.1 XP_008765851.1 3 - 4 451 65

View Full Product Details