Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Myl7
Entrez Gene ID
289759
Gene Name
myosin, light chain 7, regulatory
Gene Aliases
-
Chromosome Location
Chr.14: 86144770 - 86147075 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106017.1 NP_001099487.1 5 - 6 436 67
XM_006251216.2 XP_006251278.1 6 - 7 536 67
GenBank mRNA BC161995.1 - - 436 67

View Full Product Details