Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tcn2
Entrez Gene ID
64365
Gene Name
transcobalamin 2
Gene Aliases
Tc2, Tcn2p
Chromosome Location
Chr.14: 84174139 - 84189299 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006251366.2 XP_006251428.1 1 - 2 114 133

View Full Product Details