Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tcn2
Entrez Gene ID
64365
Gene Name
transcobalamin 2
Gene Aliases
Tc2, Tcn2p
Chromosome Location
Chr.14: 84174139 - 84189299 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022534.1 NP_071979.1 2 - 3 309 73
XM_006251366.2 XP_006251428.1 3 - 4 500 73
XM_006251367.2 XP_006251429.1 3 - 4 494 73
XM_006251368.2 XP_006251430.1 3 - 4 439 73
XM_006251370.2 XP_006251432.1 3 - 4 432 73
GenBank mRNA AF054810.1 - 2 - 3 309 73

View Full Product Details