Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gpr64
Entrez Gene ID
266735
Gene Name
G protein-coupled receptor 64
Gene Aliases
He6, Re6
Chromosome Location
Chr.X: 36930186 - 37054925 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270871.1 NP_001257800.1 15 - 16 1410 89
NM_001270872.1 NP_001257801.1 14 - 15 1374 89
NM_181366.2 NP_852031.1 16 - 17 1458 89
XM_006256916.2 XP_006256978.1 16 - 17 1498 89
XM_008773154.1 XP_008771376.1 15 - 16 1838 89
XM_008773155.1 XP_008771377.1 15 - 16 1448 89
XM_008773156.1 XP_008771378.1 14 - 15 1802 89
XM_008773157.1 XP_008771379.1 14 - 15 1796 89
XM_008773158.1 XP_008771380.1 13 - 14 1760 89
XM_008773159.1 XP_008771381.1 12 - 13 1351 89
XM_008773160.1 XP_008771382.1 15 - 16 1839 89
GenBank mRNA AF538953.1 - 16 - 17 1468 89
AF538958.1 - 15 - 16 1420 89
AF538959.1 - 14 - 15 1384 89

View Full Product Details