The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270871.1 | NP_001257800.1 | 25 - 26 | 2862 | 103 |
NM_001270872.1 | NP_001257801.1 | 24 - 25 | 2826 | 103 | |
NM_181366.2 | NP_852031.1 | 26 - 27 | 2910 | 103 | |
XM_006256916.2 | XP_006256978.1 | 26 - 27 | 2950 | 103 | |
XM_008773154.1 | XP_008771376.1 | 25 - 26 | 3290 | 103 | |
XM_008773155.1 | XP_008771377.1 | 25 - 26 | 2900 | 103 | |
XM_008773156.1 | XP_008771378.1 | 24 - 25 | 3254 | 103 | |
XM_008773157.1 | XP_008771379.1 | 24 - 25 | 3248 | 103 | |
XM_008773158.1 | XP_008771380.1 | 23 - 24 | 3212 | 103 | |
XM_008773159.1 | XP_008771381.1 | 22 - 23 | 2803 | 103 | |
GenBank mRNA | AF538953.1 | - | 26 - 27 | 2920 | 103 |
AF538958.1 | - | 25 - 26 | 2872 | 103 | |
AF538959.1 | - | 24 - 25 | 2836 | 103 |