Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zfx
Entrez Gene ID
367832
Gene Name
zinc finger protein X-linked
Gene Aliases
RGD1562664
Chromosome Location
Chr.X: 63158368 - 63233899 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109017.1 NP_001102487.1 1 - 2 372 92

View Full Product Details