Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmem41a
Entrez Gene ID
681708
Gene Name
transmembrane protein 41a
Gene Aliases
-
Chromosome Location
Chr.11: 82734525 - 82741177 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100805.1 NP_001094275.1 4 - 5 630 71
GenBank mRNA BC161997.1 - 4 - 5 630 71

View Full Product Details