Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tprg1
Entrez Gene ID
360731
Gene Name
tumor protein p63 regulated 1
Gene Aliases
RGD1306226, Svap30, Tprg
Chromosome Location
Chr.11: 78733046 - 78861745 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108320.1 NP_001101790.1 5 - 6 859 74

View Full Product Details