Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klhl1
Entrez Gene ID
290426
Gene Name
kelch-like family member 1
Gene Aliases
-
Chromosome Location
Chr.15: 80270846 - 80713309 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106054.1 NP_001099524.1 7 - 8 2298 84
XM_008770864.1 XP_008769086.1 9 - 10 2842 84

View Full Product Details