Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bpifb3
Entrez Gene ID
499924
Gene Name
BPI fold containing family B, member 3
Gene Aliases
Lplunc3, Rya3
Chromosome Location
Chr.3: 149401890 - 149416506 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109208.1 NP_001102678.1 4 - 5 499 95

View Full Product Details