The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109208.1 | NP_001102678.1 | 9 - 10 | 1010 | 60 |
XM_006235352.2 | XP_006235414.1 | 8 - 9 | 912 | 60 | |
GenBank mRNA | X60658.1 | - | 9 - 10 | 976 | 60 |