Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Insig2
Entrez Gene ID
288985
Gene Name
insulin induced gene 2
Gene Aliases
-
Chromosome Location
Chr.13: 37264818 - 37292730 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006249667.2 XP_006249729.1 1 - 2 353 86
GenBank mRNA AY156087.1 - 1 - 2 66 86

View Full Product Details