Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prdm8
Entrez Gene ID
305198
Gene Name
PR domain containing 8
Gene Aliases
-
Chromosome Location
Chr.14: 13053449 - 13060618 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008770079.1 XP_008768301.1 1 - 2 2370 66
XM_008770080.1 XP_008768302.1 2 - 3 1962 66
XM_008770081.1 XP_008768303.1 1 - 2 2370 66
XM_008770082.1 XP_008768304.1 2 - 3 500 66

View Full Product Details