The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039101.1 | NP_001034190.1 | 10 - 11 | 1171 | 71 |
XM_006257145.2 | XP_006257207.1 | 10 - 11 | 1228 | 71 | |
XM_008773347.1 | XP_008771569.1 | 11 - 12 | 1991 | 71 | |
XM_008773348.1 | XP_008771570.1 | 12 - 13 | 1144 | 71 | |
GenBank mRNA | AY886531.1 | - | 10 - 11 | 1171 | 71 |