Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tbx22
Entrez Gene ID
302369
Gene Name
T-box 22
Gene Aliases
Tbx 22
Chromosome Location
Chr.X: 78731738 - 78782542 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106937.1 NP_001100407.1 2 - 3 214 91

View Full Product Details